Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mlec
Entrez Gene ID
569613
Gene Name
malectin
Gene Aliases
si:dkey-117k10.5
Chromosome Location
Chr.8: 39870714 - 39884359 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113618.1 NP_001107090.1 4 - 5 941 111

View Full Product Details