Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ipp
Entrez Gene ID
570768
Gene Name
intracisternal A particle-promoted polypeptide
Gene Aliases
zgc:171487
Chromosome Location
Chr.2: 2993278 - 3022655 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113621.1 NP_001107093.1 1 - 2 106 105
XM_017358910.2 XP_017214399.1 1 - 2 880 105
XM_017358919.2 XP_017214408.1 1 - 2 880 105
GenBank mRNA BC154444.1 - 1 - 2 106 105
BC171521.1 - 1 - 2 101 105

View Full Product Details