Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btr05
Entrez Gene ID
794264
Gene Name
bloodthirsty-related gene family, member 5
Gene Aliases
zgc:172119
Chromosome Location
Chr.7: 16198397 - 16205820 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021477725.1 XP_021333400.1 3 - 4 1088 129

View Full Product Details