Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
exd2
Entrez Gene ID
334455
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
fi72h12, wu:fi72h12, zgc:175195
Chromosome Location
Chr.17: 51796668 - 51818713 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114406.1 NP_001107878.1 6 - 7 1126 85
XM_005169892.3 XP_005169949.1 6 - 7 1178 85
GenBank mRNA BC068411.1 - 5 - 6 692 85
BC159253.1 - 6 - 7 1126 85

View Full Product Details