Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
exd2
Entrez Gene ID
334455
Gene Name
exonuclease 3'-5' domain containing 2
Gene Aliases
fi72h12, wu:fi72h12, zgc:175195
Chromosome Location
Chr.17: 51796668 - 51818713 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114406.1 NP_001107878.1 9 - 10 1732 74
XM_005169892.3 XP_005169949.1 9 - 10 1784 74
GenBank mRNA BC068411.1 - 8 - 9 1298 74
BC159253.1 - 9 - 10 1732 74

View Full Product Details