Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
anos1b
Entrez Gene ID
30652
Gene Name
anosmin 1b
Gene Aliases
anosmin-1b, kal1.2, kal1b
Chromosome Location
Chr.22: 38823920 - 38913871 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131379.2 NP_571454.2 12 - 13 1775 69
GenBank mRNA AF163311.1 - 12 - 13 1781 69

View Full Product Details