Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kdrl
Entrez Gene ID
796537
Gene Name
kinase insert domain receptor like
Gene Aliases
flk, flk-1, flk1, kdr, kdra, vegfr-2, vegfr2, vegfr2a, vegfr4, vegr2, wu:fk52c05
Chromosome Location
Chr.14: 2386 - 17497 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131472.1 NP_571547.1 23 - 24 3117 88
XM_009291142.3 XP_009289417.1 23 - 24 3511 88
GenBank mRNA AF180354.1 - 20 - 21 2733 88
AF487829.1 - 23 - 24 3470 88
AY056466.1 - 23 - 24 3117 88
BC129158.1 - 23 - 24 3485 88
CU458916.1 - 23 - 24 3485 88
U75995.1 - 2 - 3 189 88
U82383.1 - 3 - 4 243 88
U89515.1 - 5 - 6 546 88

Gene Symbol
LOC103909948
Entrez Gene ID
103909948
Gene Name
vascular endothelial growth factor receptor kdr-like
Gene Aliases
-
Chromosome Location
Chr.Unknown: 703 - 4112 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009298309.3 XP_009296584.3 5 - 6 626 88

View Full Product Details