Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vox
Entrez Gene ID
64807
Gene Name
ventral homeobox
Gene Aliases
vega1, zgc:136532
Chromosome Location
Chr.13: 50621929 - 50624154 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131698.1 NP_571773.1 2 - 3 533 73
GenBank mRNA AF193837.1 - 2 - 3 533 73
AF255045.1 - 2 - 3 487 73
AM422095.2 - 2 - 3 348 73
BC092695.1 - 2 - 3 552 73

View Full Product Details