Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- adssl
- Entrez Gene ID
-
192305
- Gene Name
- adenylosuccinate synthase, like
- Gene Aliases
-
adss, adss2, chunp6912, zgc:66318
- Chromosome Location
-
Chr.21: 26522638 - 26531032 on Build GRCz11
- Gene Symbol
- zgc:111868
- Entrez Gene ID
-
556628
- Gene Name
- zgc:111868
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 42364157 - 42369470 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC055595.1 |
- |
- |
334 |
75 |
View Full Product Details