Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
phf5a
Entrez Gene ID
286774
Gene Name
PHD finger protein 5A
Gene Aliases
chunp6910
Chromosome Location
Chr.12: 363845 - 367781 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173266.1 NP_775373.1 2 - 3 148 76
GenBank mRNA AY099533.1 - 2 - 3 148 76

View Full Product Details