Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glulb
Entrez Gene ID
336473
Gene Name
glutamate-ammonia ligase (glutamine synthase) b
Gene Aliases
fb11d08, gso2, hm:zehn1949, im:7153900, wu:fb11d08
Chromosome Location
Chr.6: 32195 - 37427 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182866.3 NP_878286.3 5 - 6 1053 70
GenBank mRNA BC045886.1 - 5 - 6 725 70
BC066735.1 - 5 - 6 710 70
BC164204.1 - 4 - 5 638 70
BC168499.1 - 5 - 6 725 70
EH502964.1 - 2 - 3 302 70

View Full Product Details