Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hdr
Entrez Gene ID
373093
Gene Name
hematopoietic death receptor
Gene Aliases
ZH-DR, zgc:103531
Chromosome Location
Chr.5: 27404082 - 27415083 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194391.1 NP_919372.1 9 - 10 859 120
XM_005171798.4 XP_005171855.1 9 - 10 1906 120
GenBank mRNA AF302789.1 - 9 - 10 859 120
BC085545.1 - 9 - 10 1068 120
BC164857.1 - 9 - 10 1003 120

View Full Product Details