Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stap2a
Entrez Gene ID
327172
Gene Name
signal transducing adaptor family member 2a
Gene Aliases
cb901, stap2, wu:fd15d07, zgc:63913
Chromosome Location
Chr.2: 49522178 - 49569304 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198808.1 NP_942103.1 7 - 8 730 79
XM_021478992.1 XP_021334667.1 6 - 7 1037 79
GenBank mRNA BC054569.1 - 7 - 8 730 79

View Full Product Details