The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198987.2 | NP_945338.2 | 6 - 7 | 629 | 87 |
XM_021478206.1 | XP_021333881.1 | 6 - 7 | 1081 | 87 | |
GenBank mRNA | AY292652.1 | - | 6 - 7 | 87 | |
BC162774.1 | - | 6 - 7 | 629 | 87 |