Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cyp4t8
Entrez Gene ID
387527
Gene Name
cytochrome P450, family 4, subfamily T, polypeptide 8
Gene Aliases
cb1041, cyp4
Chromosome Location
Chr.16: 237325 - 280350 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199216.2 NP_954686.2 3 - 4 417 106
GenBank mRNA BC056734.1 - 3 - 4 385 106

View Full Product Details