Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ube2d3
Entrez Gene ID
321832
Gene Name
ubiquitin-conjugating enzyme E2D 3
Gene Aliases
eff, fb36f02, wu:fb36f02, zgc:56340
Chromosome Location
Chr.13: 11746242 - 11763243 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199571.1 NP_955865.1 6 - 7 648 65
GenBank mRNA BC047863.1 - 6 - 7 648 65

View Full Product Details