Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
arl8
Entrez Gene ID
327551
Gene Name
ADP-ribosylation factor-like 8
Gene Aliases
arl8l, wu:fi17a05, zgc:55350
Chromosome Location
Chr.7: 41713921 - 41726427 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199824.1 NP_956118.1 3 - 4 515 67
GenBank mRNA BC052766.1 - 3 - 4 515 67

View Full Product Details