Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmem9
Entrez Gene ID
334151
Gene Name
transmembrane protein 9
Gene Aliases
fi33g11, wu:fi33g11, zgc:55432
Chromosome Location
Chr.8: 25761624 - 25766687 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001328536.1 NP_001315465.1 2 - 3 140 73
NM_199866.2 NP_956160.2 1 - 2 402 73
XM_005167008.4 XP_005167065.1 2 - 3 182 73
XM_009303986.3 XP_009302261.1 2 - 3 178 73
GenBank mRNA BC045338.1 - 1 - 2 348 73
BC155114.1 - 2 - 3 140 73
BC164975.1 - 1 - 2 130 73
EH445611.1 - 2 - 3 161 73

View Full Product Details