Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rcvrn2
Entrez Gene ID
335650
Gene Name
recoverin 2
Gene Aliases
fk53a03, rcv1, rcv2a, wu:fk53a03, zgc:73121
Chromosome Location
Chr.16: 433192 - 442915 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199964.1 NP_956258.1 2 - 3 573 140
GenBank mRNA BC059485.1 - 2 - 3 573 140
KT325592.1 - 2 - 3 492 140

View Full Product Details