Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sdf2
Entrez Gene ID
336879
Gene Name
stromal cell-derived factor 2
Gene Aliases
fk23f01, wu:fk23f01, zgc:66291
Chromosome Location
Chr.21: 26021426 - 26028211 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200039.1 NP_956333.1 2 - 3 409 63
GenBank mRNA BC055586.1 - 2 - 3 409 63

View Full Product Details