Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
setd3
Entrez Gene ID
337193
Gene Name
SET domain containing 3, actin histidine methyltransferase
Gene Aliases
fb01d12, wu:fb01d12, zgc:63842
Chromosome Location
Chr.17: 18031899 - 18070787 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200054.1 NP_956348.1 12 - 13 1436 66
GenBank mRNA BC055261.1 - 12 - 13 1436 66
BC155278.1 - 12 - 13 1441 66

View Full Product Details