Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tmem38b
Entrez Gene ID
393145
Gene Name
transmembrane protein 38B
Gene Aliases
TRICB, zgc:55815
Chromosome Location
Chr.21: 564911 - 589018 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200176.1 NP_956470.1 - 283 84
GenBank mRNA BC045460.1 - - 283 84

View Full Product Details