Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pias4b
Entrez Gene ID
393314
Gene Name
protein inhibitor of activated STAT, 4b
Gene Aliases
pias4l, wu:fi26h11, zgc:63923
Chromosome Location
Chr.2: 57447105 - 57476462 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242942.1 NP_001229871.1 5 - 6 634 85
NM_200343.2 NP_956637.2 5 - 6 690 85
GenBank mRNA BC053149.1 - 5 - 6 674 85
EH491696.1 - 5 - 6 718 85
EH543530.1 - 5 - 6 708 85

View Full Product Details