Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
proca
Entrez Gene ID
393327
Gene Name
protein C (inactivator of coagulation factors Va and VIIIa), a
Gene Aliases
proc, si:ch1073-188c16.3, zgc:63987
Chromosome Location
Chr.2: 5457332 - 5475921 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200356.1 NP_956650.1 3 - 4 190 90
GenBank mRNA BC053182.1 - 3 - 4 190 90
BC154334.1 - 3 - 4 179 90
BC165224.1 - 1 - 2 99 90

View Full Product Details