Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trappc4
Entrez Gene ID
393737
Gene Name
trafficking protein particle complex 4
Gene Aliases
zgc:73260
Chromosome Location
Chr.15: 9315661 - 9321940 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200764.1 NP_957058.1 2 - 3 374 97
GenBank mRNA BC059594.1 - 2 - 3 374 97

View Full Product Details