Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zeb2a
Entrez Gene ID
100003889
Gene Name
zinc finger E-box binding homeobox 2a
Gene Aliases
sb:eu412, si:dkey-52o2.2, sip1a, wu:fc92g08
Chromosome Location
Chr.9: 223940 - 285859 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114551.2 NP_001108023.2 9 - 10 3211 95
XM_021478582.1 XP_021334257.1 9 - 10 3508 95
XM_021478583.1 XP_021334258.1 8 - 9 3454 95
GenBank mRNA EU379558.1 - 8 - 9 3022 95

View Full Product Details