Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gsdf
Entrez Gene ID
571289
Gene Name
gonadal somatic cell derived factor
Gene Aliases
-
Chromosome Location
Chr.21: 196490 - 205221 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114668.1 NP_001108140.1 4 - 5 503 75
XM_005173843.4 XP_005173900.1 4 - 5 1243 75
XM_017352819.1 XP_017208308.1 5 - 6 580 75
GenBank mRNA EU378916.1 - 4 - 5 503 75
JF912881.1 - 4 - 5 471 75
JF912882.1 - 4 - 5 468 75
JQ182642.1 - 4 - 5 365 75

View Full Product Details