Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
irgf1
Entrez Gene ID
100137101
Gene Name
immunity-related GTPase family, f1
Gene Aliases
-
Chromosome Location
Chr.19: 23296613 - 23300048 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320423.1 NP_001307352.1 2 - 3 707 85
XM_021467975.1 XP_021323650.1 1 - 2 750 85
GenBank mRNA BC155604.1 - 2 - 3 705 85
EH505681.1 - 2 - 3 716 85

View Full Product Details