Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdc25b
Entrez Gene ID
57925
Gene Name
cell division cycle 25B
Gene Aliases
cdc25, cdc25A, sb:cb317
Chromosome Location
Chr.13: 14915332 - 14929161 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115095.2 NP_001108567.1 4 - 5 702 84
GenBank mRNA EU921896.1 - 4 - 5 683 84

View Full Product Details