Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gapdh
Entrez Gene ID
317743
Gene Name
glyceraldehyde-3-phosphate dehydrogenase
Gene Aliases
bb02e05, cb609, gapd, mg:bb02e05, wu:fb33a10, wu:ft80f05
Chromosome Location
Chr.16: 17192671 - 17197601 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115114.1 NP_001108586.1 5 - 6 384 139
GenBank mRNA BC083506.1 - 5 - 6 384 139
BC095386.1 - 5 - 6 384 139
BC115131.1 - 5 - 6 388 139

View Full Product Details