Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ewsr1a
Entrez Gene ID
323597
Gene Name
EWS RNA-binding protein 1a
Gene Aliases
EWSR1, fc04c01, wu:fc04c01
Chromosome Location
Chr.10: 140953 - 153780 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001115138.2 NP_001108610.1 5 - 6 474 115
XM_021479109.1 XP_021334784.1 5 - 6 473 115
XM_021479110.1 XP_021334785.1 5 - 6 564 115
GenBank mRNA BC056281.1 - 5 - 6 451 115
BC068397.1 - 4 - 5 443 115
CN835703.1 - 5 - 6 485 115
FP154885.1 - 5 - 6 510 115

View Full Product Details