Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
qtrt2
Entrez Gene ID
402798
Gene Name
queuine tRNA-ribosyltransferase accessory subunit 2
Gene Aliases
qtrtd1
Chromosome Location
Chr.24: 21059673 - 21090517 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC045519.1 - 3 - 4 337 129

View Full Product Details