Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmtopsa
Entrez Gene ID
359832
Gene Name
teleost multiple tissue opsin a
Gene Aliases
tmt3a, tmtops
Chromosome Location
Chr.2: 4566196 - 4642407 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF349947.1 - - 530 60
KT008411.1 - - 698 60

View Full Product Details