Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lrp11
Entrez Gene ID
569216
Gene Name
low density lipoprotein receptor-related protein 11
Gene Aliases
si:dkey-181i3.7, wu:fj36e03
Chromosome Location
Chr.20: 1278184 - 1288059 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122874.1 NP_001116346.1 5 - 6 1681 86

View Full Product Details