Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tnn
Entrez Gene ID
30234
Gene Name
tenascin N
Gene Aliases
tenw, tnw, zgc:110729
Chromosome Location
Chr.2: 35509418 - 35567016 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131036.2 NP_571111.2 4 - 5 1188 97
XM_005171264.4 XP_005171321.1 4 - 5 1183 97
XM_005171265.3 XP_005171322.1 4 - 5 1183 97
XM_009298633.3 XP_009296908.1 4 - 5 1183 97
GenBank mRNA AJ001423.1 - 4 - 5 1148 97
BC071284.1 - 4 - 5 1151 97
BC095389.1 - 4 - 5 1110 97
KM655642.1 - 3 - 4 1022 97

View Full Product Details