Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
th
Entrez Gene ID
30384
Gene Name
tyrosine hydroxylase
Gene Aliases
-
Chromosome Location
Chr.25: 24055341 - 24074500 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131149.1 NP_571224.1 2 - 3 284 64
GenBank mRNA BC163630.1 - 2 - 3 351 64
BC163639.1 - 2 - 3 351 64

View Full Product Details