Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
derl3
Entrez Gene ID
393877
Gene Name
derlin 3
Gene Aliases
zgc:63991
Chromosome Location
Chr.8: 154093 - 162333 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC064666.1 - 6 - 7 823 107

View Full Product Details