Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rab13
Entrez Gene ID
373105
Gene Name
RAB13, member RAS oncogene family
Gene Aliases
cb764, hm:zeh0455
Chromosome Location
Chr.16: 23799745 - 23809611 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201329.1 NP_958486.1 5 - 6 810 88
GenBank mRNA BC053195.1 - 5 - 6 810 88

View Full Product Details