Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnaja3b
Entrez Gene ID
394242
Gene Name
DnaJ heat shock protein family (Hsp40) member A3b
Gene Aliases
si:dkey-30o19.2
Chromosome Location
Chr.22: 25931837 - 25943267 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201342.1 NP_958499.1 1 - 2 194 96
GenBank mRNA BC055555.1 - 1 - 2 194 96

Gene Symbol
LOC554962
Entrez Gene ID
554962
Gene Name
J domain-containing protein
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123236.1 NP_001116708.1 1 - 2 194 96

View Full Product Details