Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aldh9a1a.1
Entrez Gene ID
100005587
Gene Name
aldehyde dehydrogenase 9 family, member A1a, tandem duplicate 1
Gene Aliases
TMABA-DH, TMABADH, aldh9a1a, aldh9a1l, wu:fb64d09
Chromosome Location
Chr.8: 129586 - 139327 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201471.1 NP_958879.1 3 - 4 369 78
XM_005166774.3 XP_005166831.1 3 - 4 474 78
GenBank mRNA BC045932.1 - 3 - 4 369 78
BC066668.1 - 3 - 4 362 78
BC165753.1 - 2 - 3 332 78
BC166038.1 - 3 - 4 371 78
BC167472.1 - 3 - 4 371 78

View Full Product Details