Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aldh9a1a.1
Entrez Gene ID
100005587
Gene Name
aldehyde dehydrogenase 9 family, member A1a, tandem duplicate 1
Gene Aliases
TMABA-DH, TMABADH, aldh9a1a, aldh9a1l, wu:fb64d09
Chromosome Location
Chr.8: 129586 - 139327 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201471.1 NP_958879.1 6 - 7 823 77
XM_005166774.3 XP_005166831.1 6 - 7 928 77
GenBank mRNA BC045932.1 - 6 - 7 823 77
BC066668.1 - 6 - 7 816 77
BC165753.1 - 5 - 6 786 77
BC166038.1 - 6 - 7 825 77
BC167472.1 - 6 - 7 825 77

View Full Product Details