Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mmp13a
Entrez Gene ID
387293
Gene Name
matrix metallopeptidase 13a
Gene Aliases
cb1034, mmp13
Chromosome Location
Chr.10: 38593657 - 38619868 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC049472.1 - 2 - 3 390 85

View Full Product Details