Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mapk3
Entrez Gene ID
399480
Gene Name
mitogen-activated protein kinase 3
Gene Aliases
ERK1, fi06b09, wu:fi06b09, zERK1
Chromosome Location
Chr.3: 26211974 - 26222308 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201507.1 NP_958915.1 1 - 2 372 124
GenBank mRNA AB030902.1 - 1 - 2 291 124
AY922319.1 - 1 - 2 210 124
BC045505.1 - 1 - 2 372 124
BC066401.1 - 1 - 2 360 124
BC097073.1 - 1 - 2 445 124
BC163960.1 - 1 - 2 244 124

View Full Product Details