Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mapk3
Entrez Gene ID
399480
Gene Name
mitogen-activated protein kinase 3
Gene Aliases
ERK1, fi06b09, wu:fi06b09, zERK1
Chromosome Location
Chr.3: 26211974 - 26222308 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201507.1 NP_958915.1 7 - 8 1223 63
GenBank mRNA AY922319.1 - 7 - 8 1061 63
BC045505.1 - 7 - 8 1223 63
BC066401.1 - 7 - 8 1211 63
BC097073.1 - 7 - 8 1296 63
BC163960.1 - 7 - 8 1095 63

View Full Product Details