Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
micall2a
Entrez Gene ID
327198
Gene Name
mical-like 2a
Gene Aliases
fd16d06, micall2, wu:fd16d06, wu:fd16d07, zgc:55358
Chromosome Location
Chr.3: 43102010 - 43135493 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001327787.1 NP_001314716.1 13 - 14 2319 66
NR_137174.1 14 - 15 2504 66
XM_005164196.4 XP_005164253.1 13 - 14 2328 66
XM_005164197.4 XP_005164254.1 13 - 14 2325 66
XM_005164198.4 XP_005164255.1 13 - 14 2322 66
GenBank mRNA BC052767.1 - 14 - 15 2406 66
HM151009.1 - 13 - 14 2081 66

View Full Product Details