Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
micall2a
Entrez Gene ID
327198
Gene Name
mical-like 2a
Gene Aliases
fd16d06, micall2, wu:fd16d06, wu:fd16d07, zgc:55358
Chromosome Location
Chr.3: 43102010 - 43135493 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001327787.1 NP_001314716.1 14 - 15 2384 107
NR_137174.1 15 - 16 2569 107
XM_005164196.4 XP_005164253.1 14 - 15 2393 107
XM_005164197.4 XP_005164254.1 14 - 15 2390 107
XM_005164198.4 XP_005164255.1 14 - 15 2387 107
GenBank mRNA BC052767.1 - 15 - 16 107
HM151009.1 - 14 - 15 107

View Full Product Details