Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smyd1a
Entrez Gene ID
321245
Gene Name
SET and MYND domain containing 1a
Gene Aliases
sb:cb405, smyd1, wu:fc09a05, zgc:77936
Chromosome Location
Chr.5: 72111189 - 72125515 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205540.2 NP_991103.2 6 - 7 897 91
GenBank mRNA BC065475.1 - 6 - 7 897 91
EH601391.1 - 4 - 5 639 91

View Full Product Details