Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
hnf4b
Entrez Gene ID
324010
Gene Name
hepatic nuclear factor 4, beta
Gene Aliases
fc16g07, wu:fc16g07, zgc:77387
Chromosome Location
Chr.7: 69019595 - 69047681 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205546.1 NP_991109.1 - 1526 75
GenBank mRNA BC065657.1 - - 1526 75

View Full Product Details