Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hey1
Entrez Gene ID
58008
Gene Name
hes-related family bHLH transcription factor with YRPW motif 1
Gene Aliases
id:ibd1292, zgc:110572
Chromosome Location
Chr.19: 31797567 - 31802275 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212561.1 NP_997726.1 1 - 2 130 87
GenBank mRNA AJ510221.1 - 1 - 2 130 87
BC095317.1 - 1 - 2 151 87
BC164111.1 - 1 - 2 119 87

View Full Product Details