Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mcm7
Entrez Gene ID
192333
Gene Name
minichromosome maintenance complex component 7
Gene Aliases
chunp6911, nyz175, sr:nyz175
Chromosome Location
Chr.14: 93967 - 102282 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212569.1 NP_997734.1 - 633 105
GenBank mRNA AY099527.1 - - 494 105
BC045497.1 - - 633 105
BC065669.1 - - 485 105
MH746781.1 - - 553 105

View Full Product Details